siRNA Details
| Gene Symbol: Frat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177603.3 | NP_808271.1 | 1 | 1984 | ||
| GenBank mRNA | AK081171.1 | - | 1 | 1985 | ||
| BC034214.1 | - | 1 | 1947 | 4989784 | ||
| BC085510.1 | - | 1 | 1960 | 5718518 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frat2| Gene Symbol: Frat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177603.3 | NP_808271.1 | 1 | 1984 | ||
| GenBank mRNA | AK081171.1 | - | 1 | 1985 | ||
| BC034214.1 | - | 1 | 1947 | 4989784 | ||
| BC085510.1 | - | 1 | 1960 | 5718518 | ||