siRNA Details
| Gene Symbol: AA986860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177604.3 | NP_808272.2 | 4 | 2759 | ||
| GenBank mRNA | AK028957.1 | - | 4 | 2651 | ||
| AK085225.1 | - | 2 | 3054 | |||
| AK086121.1 | - | 4 | 2635 | |||
| BC099401.1 | - | 1 | 1698 | 30713499 | ||

AA986860
Genomic Map