siRNA Details
| Gene Symbol: Wscd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177618.4 | NP_808286.2 | 2, 3 | 810 | ||
| GenBank mRNA | AK220346.1 | - | 1, 2 | 516 | ||
| BC030477.1 | - | 2, 3 | 948 | |||
| BC048831.1 | - | 1, 2 | 629 | 4504495 | ||
| BC063070.1 | - | 2, 3 | 810 | 5687975 | ||

Wscd1
Genomic Map