siRNA Details
| Gene Symbol: Rin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161365.1 | NP_001154837.1 | 7 | 2239 | ||
| NM_177620.4 | NP_808288.2 | 7 | 2350 | |||
| GenBank mRNA | AK032205.1 | - | 7 | 2351 | ||
| AK154506.1 | - | 7 | 2375 | |||
| AK154963.1 | - | 7 | 2240 | |||
| AK171301.1 | - | 7 | 2164 | |||
| AK202699.1 | - | 1 | 35 | |||
| AK206610.1 | - | 1 | 35 | |||
| AY265356.1 | - | 7 | 2336 | |||
| BC137907.1 | - | 7 | 2278 | 8860926 | ||

Rin3
Genomic Map