siRNA Details
| Gene Symbol: H2-M10.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177637.3 | NP_808305.2 | 3 | 491 | ||
| GenBank mRNA | AF539601.1 | - | 3 | 491 | ||
| AY211082.1 | - | 3 | 500 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2-M10.5| Gene Symbol: H2-M10.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177637.3 | NP_808305.2 | 3 | 491 | ||
| GenBank mRNA | AF539601.1 | - | 3 | 491 | ||
| AY211082.1 | - | 3 | 500 | |||