siRNA Details
| Gene Symbol: Hmcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083701.1 | - | 10 | 1350 | ||
| BC034076.1 | - | 11 | 1780 | |||
| BC040753.1 | - | 11 | 1780 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmcn2| Gene Symbol: Hmcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083701.1 | - | 10 | 1350 | ||
| BC034076.1 | - | 11 | 1780 | |||
| BC040753.1 | - | 11 | 1780 | |||