siRNA Details
| Gene Symbol: BC034902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040732.1 | - | 2 | 631 | ||
| BC034902.1 | - | 2 | 542 | 1364939 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC034902| Gene Symbol: BC034902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040732.1 | - | 2 | 631 | ||
| BC034902.1 | - | 2 | 542 | 1364939 | ||