siRNA Details
| Gene Symbol: Isg20l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177663.4 | NP_808331.1 | 2 | 988 | ||
| GenBank mRNA | AK053778.1 | - | 2 | 989 | ||
| AK076747.1 | - | 2 | 710 | |||
| AK155979.1 | - | 2 | 992 | |||
| AK178586.1 | - | 1 | 280 | |||
| BC098326.1 | - | 2 | 975 | 5720443 | ||

Isg20l2
Genomic Map