siRNA Details
| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122730.2 | NP_001116202.2 | 7 | 2720 | ||
| NM_178242.2 | NP_839973.1 | 7 | 2720 | |||
| GenBank mRNA | AK173280.1 | - | 4 | 475 | ||
| BC025631.1 | - | 2 | 250 | |||
| BC049818.1 | - | 7 | 2720 | 6511384 | ||
| BC115863.1 | - | 3 | 366 | |||

Tnrc18
Genomic Map