siRNA Details
| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178242.2 | NP_839973.1 | 16 | 5874 | ||
| GenBank mRNA | BC049818.1 | - | 16 | 5874 | 6511384 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnrc18| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178242.2 | NP_839973.1 | 16 | 5874 | ||
| GenBank mRNA | BC049818.1 | - | 16 | 5874 | 6511384 | |