siRNA Details
| Gene Symbol: Rcsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038846.1 | NP_001033935.1 | 5 | 1185 | ||
| NM_178593.3 | NP_848708.2 | 6 | 1275 | |||
| GenBank mRNA | AK133961.1 | - | 6 | 1230 | ||
| AK158429.1 | - | 6 | 1216 | |||
| AK168816.1 | - | 5 | 1156 | |||
| AK171323.1 | - | 1165 | ||||
| AK171914.1 | - | 6 | 1198 | |||
| BC025872.1 | - | 6 | 1195 | 5037953 | ||

Rcsd1
Genomic Map