siRNA Details
| Gene Symbol: Tfdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184706.1 | NP_001171635.1 | 7, 8 | 886 | ||
| NM_001184708.1 | NP_001171637.1 | 4, 5 | 612 | |||
| NM_001184709.1 | NP_001171638.1 | 5, 6 | 510 | |||
| NM_001184710.1 | NP_001171639.1 | 4, 5 | 386 | |||
| NM_178667.4 | NP_848782.2 | 8, 9 | 887 | |||
| GenBank mRNA | AK029055.1 | - | 7, 8 | 859 | ||
| AK033356.1 | - | 5, 6 | 474 | |||
| AK044784.1 | - | 4, 5 | 614 | |||
| AK053881.1 | - | 7, 8 | 845 | |||
| AK079624.1 | - | 8, 9 | 815 | |||
| AK158829.1 | - | 4, 5 | 382 | |||
| BC141080.1 | - | 6, 7 | 544 | 9055692 | ||

Tfdp2
Genomic Map