siRNA Details
| Gene Symbol: Pde12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178668.3 | NP_848783.3 | 2, 3 | 1686 | ||
| GenBank mRNA | AK051991.1 | - | 902 | |||
| AK082947.1 | - | 2, 3 | 1509 | |||
| AK088843.1 | - | 2, 3 | 1499 | |||
| AK143567.1 | - | 2, 3 | 1513 | |||
| AK155088.1 | - | 1520 | ||||
| AK161083.1 | - | 2, 3 | 1726 | |||
| AK167708.1 | - | 2, 3 | 1484 | |||
| AK201375.1 | - | 238 | ||||
| AK215000.1 | - | 2, 3 | 237 | |||
| BC064450.1 | - | 2, 3 | 1686 | 6417852 | ||

Pde12
Genomic Map