siRNA Details
| Gene Symbol: Cd226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039149.1 | NP_001034238.1 | 3 | 528 | ||
| NM_178687.2 | NP_848802.2 | 4 | 867 | |||
| GenBank mRNA | AB195429.1 | - | 3 | 640 | ||
| AF416980.1 | - | 4 | 699 | |||
| AF421199.1 | - | 3 | 360 | |||
| AF421200.1 | - | 2 | 288 | |||
| AK036455.1 | - | 5 | 962 | |||
| AK140349.1 | - | 4 | 755 | |||
| BC051526.1 | - | 3 | 462 | |||
| BC132422.1 | - | 4 | 812 | 40130699 | ||
| BC138769.1 | - | 4 | 812 | 8861791 | ||
| BC145410.1 | - | 3 | 473 | 9053968 | ||

Cd226
Genomic Map