siRNA Details
| Gene Symbol: CRISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190986.1 | NP_001177915.1 | 3 | 266 | ||
| NM_006061.2 | NP_006052.2 | 3 | 254 | |||
| GenBank mRNA | AK223380.1 | - | 3 | 158 | ||
| AK292786.1 | - | 3 | 213 | |||
| AK313387.1 | - | 3 | 187 | |||
| BC069580.1 | - | 2 | 140 | 7262396 | ||
| BC069602.1 | - | 2 | 142 | 7262408 | ||
| BC101539.1 | - | 2 | 164 | 8069045 | ||
| DB210604.1 | - | 3 | 254 | |||
| X94323.1 | - | 3 | 167 | |||
| X95240.1 | - | 2 | 142 | |||

CRISP3
Genomic Map