siRNA Details
| Gene Symbol: Pigv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145955.1 | NP_001139427.1 | 3, 4 | 736 | ||
| NM_001145956.1 | NP_001139428.1 | 2, 3 | 460 | |||
| NM_178698.5 | NP_848813.3 | 3, 4 | 1582 | |||
| GenBank mRNA | AK046527.1 | - | 3, 4 | 1543 | ||
| AK047322.1 | - | 3, 4 | 1380 | |||
| AK052446.1 | - | 3, 4 | 1556 | |||
| AK080590.1 | - | 2, 3 | 461 | |||
| AK080858.1 | - | 3, 4 | 1536 | |||
| AK148285.1 | - | 2, 3 | 449 | |||
| BC055060.1 | - | 3, 4 | 1532 | 6528864 | ||

Pigv
Genomic Map