siRNA Details
| Gene Symbol: Pigv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145955.1 | NP_001139427.1 | 4 | 2067 | ||
| NM_001145956.1 | NP_001139428.1 | 3 | 1791 | |||
| NM_178698.5 | NP_848813.3 | 4 | 2913 | |||
| GenBank mRNA | AK052446.1 | - | 4 | 2886 | ||
| AK080858.1 | - | 4 | 2867 | |||
| AK148285.1 | - | 3 | 1779 | |||

Pigv
Genomic Map