siRNA Details
| Gene Symbol: B930041F14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178699.4 | NP_848814.3 | 1 | 1208 | ||
| GenBank mRNA | AB041660.1 | - | 1 | 729 | ||
| AK046537.1 | - | 1 | 1211 | |||
| AK047240.1 | - | 1 | 1079 | |||
| AK048841.1 | - | 1 | 1053 | |||

B930041F14Rik
Genomic Map