siRNA Details
| Gene Symbol: Lrrc8d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122768.1 | NP_001116240.1 | 3 | 3172 | ||
| NM_178701.3 | NP_848816.3 | 3 | 3162 | |||
| GenBank mRNA | AK046249.1 | - | 3 | 3171 | ||
| AK050400.1 | - | 3 | 3132 | |||
| AK080722.1 | - | 3 | 3106 | |||
| AK137459.1 | - | 3 | 3173 | |||
| AK167985.1 | - | 2 | 2880 | |||
| AK171882.1 | - | 1 | 72 | |||
| AK207105.1 | - | 1 | 135 | |||
| BC014756.1 | - | 1 | 2196 | |||
| BC037717.1 | - | 3 | 3143 | 4239389 | ||

Lrrc8d
Genomic Map