siRNA Details
| Gene Symbol: Dpy19l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178704.3 | NP_848819.2 | 11 | 1384 | ||
| GenBank mRNA | AK031345.1 | - | 11 | 1376 | ||
| AK033395.1 | - | 11 | 1341 | |||
| AK034217.1 | - | 11 | 1385 | |||
| AK045758.1 | - | 11 | 1285 | |||
| BC050065.1 | - | 11 | 1328 | 30016211 | ||
| BC059897.1 | - | 11 | 1281 | 6821433 | ||

Dpy19l3
Genomic Map