siRNA Details
| Gene Symbol: EMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142288.1 | NP_001135760.1 | 2, 3 | 555 | ||
| NM_006067.4 | NP_006058.1 | 2, 3 | 555 | |||
| XM_017022867.1 | XP_016878356.1 | 2, 3 | 246 | |||
| GenBank mRNA | AF005888.1 | - | 2, 3 | 424 | ||
| AK313977.1 | - | 2, 3 | 537 | |||
| BC001472.2 | - | 2, 3 | 507 | 3138396 | ||
| BC005886.2 | - | 2, 3 | 507 | 3546377 | ||
| BC007445.1 | - | 2, 3 | 498 | 3009805 | ||
| BC020250.1 | - | 2, 3 | 507 | 4653699 | ||
| BE855452.1 | - | 2, 3 | 429 | |||
| BP307869.1 | - | 2, 3 | 560 | |||
| BT007132.1 | - | 2, 3 | 308 | |||

EMC8
Genomic Map