siRNA Details
| Gene Symbol: Rfesd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131068.1 | NP_001124540.1 | 4 | 970 | ||
| NM_001131069.1 | NP_001124541.1 | 4 | 433 | |||
| NM_178916.5 | NP_849247.3 | 4 | 1420 | |||
| GenBank mRNA | AK029602.1 | - | 4 | 1246 | ||
| AK040255.1 | - | 4 | 925 | |||
| AK083705.1 | - | 4 | 1421 | |||
| BC030381.1 | - | 4 | 1045 | 5375753 | ||
| CN839985.1 | - | 4 | 443 | |||

Rfesd
Genomic Map