siRNA Details
| Gene Symbol: TRDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006073.3 | NP_006064.2 | 23 | 1758 | ||
| GenBank mRNA | CR749591.1 | - | 12 | 466 | ||
| U18985.1 | - | 23 | 1462 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRDN| Gene Symbol: TRDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006073.3 | NP_006064.2 | 23 | 1758 | ||
| GenBank mRNA | CR749591.1 | - | 12 | 466 | ||
| U18985.1 | - | 23 | 1462 | |||