siRNA Details
| Gene Symbol: Dsg1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181682.1 | NP_859010.1 | 15 | 3351 | ||
| GenBank mRNA | AK036986.1 | - | 15 | 3458 | ||
| AY192158.1 | - | 15 | 3351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dsg1b| Gene Symbol: Dsg1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181682.1 | NP_859010.1 | 15 | 3351 | ||
| GenBank mRNA | AK036986.1 | - | 15 | 3458 | ||
| AY192158.1 | - | 15 | 3351 | |||