siRNA Details
| Gene Symbol: Gpr142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181749.1 | NP_861414.1 | 1, 2 | 102 | ||
| GenBank mRNA | AY288428.1 | - | 1, 2 | 102 | ||
| AY957501.1 | - | 1, 2 | 102 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr142| Gene Symbol: Gpr142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181749.1 | NP_861414.1 | 1, 2 | 102 | ||
| GenBank mRNA | AY288428.1 | - | 1, 2 | 102 | ||
| AY957501.1 | - | 1, 2 | 102 | |||