siRNA Details
| Gene Symbol: Fam167b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182783.2 | NP_877584.2 | 2 | 705 | ||
| GenBank mRNA | AK134621.1 | - | 2 | 535 | ||
| AK143928.1 | - | 2 | 675 | |||
| BC030183.1 | - | 2 | 672 | |||
| BX517339.1 | - | 2 | 288 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam167b| Gene Symbol: Fam167b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182783.2 | NP_877584.2 | 2 | 705 | ||
| GenBank mRNA | AK134621.1 | - | 2 | 535 | ||
| AK143928.1 | - | 2 | 675 | |||
| BC030183.1 | - | 2 | 672 | |||
| BX517339.1 | - | 2 | 288 | |||