siRNA Details
Gene Symbol: Iqsec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134383.1 | NP_001127855.1 | 14 | 3536 | ||
NM_001134384.1 | NP_001127856.1 | 14 | 5500 | |||
GenBank mRNA | AK044711.1 | - | 1 | 1701 | ||
BC026481.1 | - | 12 | 2040 | |||
BC043918.1 | - | 1 | 512 | |||
BC096455.1 | - | 1 | 1169 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
