siRNA Details
| Gene Symbol: Iffo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205173.1 | NP_001192102.1 | 5 | 1250 | ||
| NM_183148.3 | NP_898971.1 | 8 | 870 | |||
| GenBank mRNA | BC027170.1 | - | 4 | 396 | 4952810 | |
| BC100381.1 | - | 8 | 870 | |||
| BE860309.1 | - | 3 | 239 | |||
| CN718972.1 | - | 2 | 214 | |||

Iffo2
Genomic Map