siRNA Details
| Gene Symbol: AI987944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199330.1 | NP_001186259.1 | 5 | 789 | ||
| NM_183167.4 | NP_898990.1 | 4 | 522 | |||
| GenBank mRNA | BC052842.1 | - | 4 | 534 | 30051444 | |
| BC099396.1 | - | 5 | 789 | 30939449 | ||
| BC138015.1 | - | 5 | 670 | 8861034 | ||

AI987944
Genomic Map