siRNA Details
| Gene Symbol: Sbno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183426.1 | NP_906271.1 | 5, 6 | 625 | ||
| GenBank mRNA | AJ512611.1 | - | 6, 7 | 781 | ||
| AK089496.1 | - | 2, 3 | 370 | |||
| AK149806.1 | - | 5, 6 | 649 | |||
| AK155462.1 | - | 5, 6 | 623 | |||
| AK155674.1 | - | 5, 6 | 622 | |||
| BC056369.1 | - | 5, 6 | 625 | 6831600 | ||
| BC064113.1 | - | 2, 3 | 382 | |||

Sbno2
Genomic Map