siRNA Details
| Gene Symbol: Sbno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183426.1 | NP_906271.1 | 9 | 991 | ||
| GenBank mRNA | AJ512611.1 | - | 10 | 1147 | ||
| AK089496.1 | - | 5 | 910 | |||
| AK149806.1 | - | 9 | 1015 | |||
| AK155462.1 | - | 9 | 989 | |||
| AK155674.1 | - | 9 | 988 | |||
| AK170733.1 | - | 8 | 783 | |||
| BC056369.1 | - | 9 | 991 | 6831600 | ||
| BC064113.1 | - | 6 | 748 | |||

Sbno2
Genomic Map