siRNA Details
| Gene Symbol: Synrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194341.2 | NP_919322.1 | 14 | 2627 | ||
| GenBank mRNA | AK142858.1 | - | 1 | 625 | ||
| BC056370.1 | - | 14 | 2587 | 6831893 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Synrg| Gene Symbol: Synrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194341.2 | NP_919322.1 | 14 | 2627 | ||
| GenBank mRNA | AK142858.1 | - | 1 | 625 | ||
| BC056370.1 | - | 14 | 2587 | 6831893 | ||