siRNA Details
| Gene Symbol: Alkbh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198027.2 | NP_932144.2 | 5, 6 | 442 | ||
| GenBank mRNA | AK209174.1 | - | 1, 2 | 90 | ||
| BC029805.1 | - | 5, 6 | 394 | 4952089 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alkbh6| Gene Symbol: Alkbh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198027.2 | NP_932144.2 | 5, 6 | 442 | ||
| GenBank mRNA | AK209174.1 | - | 1, 2 | 90 | ||
| BC029805.1 | - | 5, 6 | 394 | 4952089 | ||