siRNA Details
| Gene Symbol: Med16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163276.1 | NP_001156748.1 | 2, 3 | 291 | ||
| NM_198107.2 | NP_932775.2 | 2, 3 | 294 | |||
| GenBank mRNA | AK042340.1 | - | 2, 3 | 256 | ||
| AK138125.1 | - | 2, 3 | 278 | |||
| AK149345.1 | - | 2, 3 | 275 | |||
| AK196235.1 | - | 1, 2 | 98 | |||
| BC057056.1 | - | 2, 3 | 248 | 6813010 | ||
| BY019373.1 | - | 2, 3 | 295 | |||

Med16
Genomic Map