siRNA Details
| Gene Symbol: Tmem63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198167.3 | NP_937810.2 | 10 | 851 | ||
| GenBank mRNA | AK090200.1 | - | 10 | 772 | ||
| AK159673.1 | - | 10 | 856 | |||
| BC059283.1 | - | 3 | 199 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem63b| Gene Symbol: Tmem63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198167.3 | NP_937810.2 | 10 | 851 | ||
| GenBank mRNA | AK090200.1 | - | 10 | 772 | ||
| AK159673.1 | - | 10 | 856 | |||
| BC059283.1 | - | 3 | 199 | |||