siRNA Details
| Gene Symbol: Gmeb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198169.2 | NP_937812.1 | 6, 7 | 746 | ||
| GenBank mRNA | AB221583.1 | - | 5, 6 | 626 | ||
| AB221584.1 | - | 5, 6 | 633 | |||
| AK129319.1 | - | 2, 3 | 278 | |||
| BC029763.1 | - | 5, 6 | 569 | |||
| BC059066.1 | - | 6, 7 | 746 | 6825428 | ||

Gmeb2
Genomic Map