siRNA Details
| Gene Symbol: Gmeb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198169.2 | NP_937812.1 | 10 | 3371 | ||
| GenBank mRNA | AK129319.1 | - | 5 | 3621 | ||
| BC029763.1 | - | 9 | 3194 | |||
| BC059066.1 | - | 10 | 3371 | 6825428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmeb2| Gene Symbol: Gmeb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198169.2 | NP_937812.1 | 10 | 3371 | ||
| GenBank mRNA | AK129319.1 | - | 5 | 3621 | ||
| BC029763.1 | - | 9 | 3194 | |||
| BC059066.1 | - | 10 | 3371 | 6825428 | ||