siRNA Details
| Gene Symbol: Rbm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198302.2 | NP_938044.1 | 5 | 525 | ||
| GenBank mRNA | AK039297.1 | - | 5 | 467 | ||
| BC050779.1 | - | 5 | 572 | 6771154 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm11| Gene Symbol: Rbm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198302.2 | NP_938044.1 | 5 | 525 | ||
| GenBank mRNA | AK039297.1 | - | 5 | 467 | ||
| BC050779.1 | - | 5 | 572 | 6771154 | ||