siRNA Details
| Gene Symbol: Klhl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198305.2 | NP_938047.1 | 5, 6 | 919 | ||
| GenBank mRNA | AK049214.1 | - | 5, 6 | 796 | ||
| AK154523.1 | - | 5, 6 | 857 | |||
| AK170825.1 | - | 5, 6 | 857 | |||
| AY423764.1 | - | 5, 6 | 897 | |||
| BC139309.1 | - | 5, 6 | 860 | 8862331 | ||

Klhl17
Genomic Map