siRNA Details
| Gene Symbol: Klhl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198305.2 | NP_938047.1 | 12 | 2227 | ||
| GenBank mRNA | AK049214.1 | - | 11 | 2357 | ||
| AY423764.1 | - | 12 | 2205 | |||
| BC024128.1 | - | 1 | 836 | |||
| BC048408.1 | - | 1 | 777 | |||
| BC058738.1 | - | 5 | 814 | |||
| BC139309.1 | - | 12 | 2168 | 8862331 | ||

Klhl17
Genomic Map