siRNA Details
| Gene Symbol: Wnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185020.1 | NP_001171949.1 | 7 | 2967 | ||
| NM_001185021.1 | NP_001171950.1 | 7 | 2967 | |||
| NM_001199083.1 | NP_001186012.1 | 7 | 2967 | |||
| NM_001199084.1 | NP_001186013.1 | 7 | 2967 | |||
| NM_198703.3 | NP_941992.2 | 7 | 2967 | |||
| GenBank mRNA | AK052468.1 | - | 5 | 797 | ||
| AK172935.1 | - | 8 | 2204 | |||
| AY309076.1 | - | 7 | 2091 | |||
| AY455867.1 | - | 7 | 2109 | |||
| BC030370.1 | - | 4 | 732 | |||
| BC138445.1 | - | 7 | 1971 | 8861465 | ||
| BC145282.1 | - | 7 | 1971 | 9053836 | ||
| BC171955.1 | - | 7 | 1971 | 9054241 | ||

Wnk1
Genomic Map