siRNA Details
| Gene Symbol: Smgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198927.3 | NP_945121.1 | 2 | 122 | ||
| GenBank mRNA | AK017450.1 | - | 2 | 123 | ||
| AK132579.1 | - | 2 | 124 | |||
| AY459348.1 | - | 2 | 123 | |||
| BC055490.1 | - | 2 | 104 | |||
| BC138091.1 | - | 2 | 123 | 8861110 | ||
| BC145136.1 | - | 2 | 123 | 9053684 | ||

Smgc
Genomic Map