siRNA Details
| Gene Symbol: Smgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198927.3 | NP_945121.1 | 8, 9 | 1007 | ||
| GenBank mRNA | AK017450.1 | - | 8, 9 | 1007 | ||
| AK132579.1 | - | 8, 9 | 1009 | |||
| AY459348.1 | - | 8, 9 | 1008 | |||
| BC049243.1 | - | 12 | ||||
| BC069958.1 | - | 2, 3 | 199 | |||
| BC138091.1 | - | 8, 9 | 1008 | 8861110 | ||
| BC145136.1 | - | 5, 6 | 648 | 9053684 | ||

Smgc
Genomic Map