siRNA Details
| Gene Symbol: Fam171a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199200.2 | NP_954670.2 | 8 | 1254 | ||
| GenBank mRNA | BC025575.1 | - | 8 | 1230 | ||
| BC064455.1 | - | 5 | 987 | 6810415 | ||
| CB202938.1 | - | 6 | 720 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam171a2| Gene Symbol: Fam171a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199200.2 | NP_954670.2 | 8 | 1254 | ||
| GenBank mRNA | BC025575.1 | - | 8 | 1230 | ||
| BC064455.1 | - | 5 | 987 | 6810415 | ||
| CB202938.1 | - | 6 | 720 | |||