siRNA Details
Gene Symbol: Fam171a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199200.2 | NP_954670.2 | 8 | 3052 | ||
GenBank mRNA | BC064455.1 | - | 6 | 2634 | 6810415 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam171a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199200.2 | NP_954670.2 | 8 | 3052 | ||
GenBank mRNA | BC064455.1 | - | 6 | 2634 | 6810415 |