siRNA Details
| Gene Symbol: Gcm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008104.2 | NP_032130.2 | 5 | 788 | ||
| GenBank mRNA | AF081556.1 | - | 5 | 754 | ||
| AK132499.1 | - | 3 | 572 | |||
| AK135689.1 | - | 7 | 787 | |||
| BC110631.1 | - | 5 | 818 | 40054293 | ||
| BC110632.1 | - | 5 | 818 | 40054295 | ||
| D88611.1 | - | 5 | 995 | |||

Gcm2
Genomic Map