siRNA Details
| Gene Symbol: Kif15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010620.1 | NP_034750.1 | 3, 4 | 307 | ||
| GenBank mRNA | AJ560623.1 | - | 3, 4 | 324 | ||
| AK050821.1 | - | 3, 4 | 324 | |||
| BC060710.1 | - | 3, 4 | 307 | 6409082 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif15| Gene Symbol: Kif15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010620.1 | NP_034750.1 | 3, 4 | 307 | ||
| GenBank mRNA | AJ560623.1 | - | 3, 4 | 324 | ||
| AK050821.1 | - | 3, 4 | 324 | |||
| BC060710.1 | - | 3, 4 | 307 | 6409082 | ||