siRNA Details
| Gene Symbol: Slc35b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016752.1 | NP_058032.1 | 3, 4 | 435 | ||
| GenBank mRNA | AK002961.1 | - | 3, 4 | 429 | ||
| AK007711.1 | - | 3, 4 | 441 | |||
| AK160124.1 | - | 3, 4 | 501 | |||
| AK183623.1 | - | 2, 3 | 219 | |||
| AK184790.1 | - | 2, 3 | 219 | |||
| AK185566.1 | - | 2, 3 | 216 | |||
| BC002029.1 | - | 3, 4 | 416 | 3592141 | ||
| D87990.1 | - | 3, 4 | 435 | |||

Slc35b1
Genomic Map