siRNA Details
| Gene Symbol: Caln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021371.2 | NP_067346.1 | 6 | 1426 | ||
| NM_181045.1 | NP_851388.1 | 7 | 1126 | |||
| GenBank mRNA | AB041539.1 | - | 6 | 1417 | ||
| AF282251.1 | - | 7 | 1108 | |||
| AK035865.1 | - | 7 | 1095 | |||
| AK081011.1 | - | 6 | 860 | |||
| AK158641.1 | - | 6 | 788 | |||
| AK158892.1 | - | 6 | 1398 | |||
| AK163648.1 | - | 6 | 889 | |||
| BC057129.1 | - | 6 | 1405 | 6811387 | ||
| BC058963.1 | - | 6 | 1405 | 6811387 | ||

Caln1
Genomic Map