siRNA Details
| Gene Symbol: Caln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021371.2 | NP_067346.1 | 6 | 1615 | ||
| NM_181045.1 | NP_851388.1 | 7 | 1315 | |||
| GenBank mRNA | AB041539.1 | - | 6 | 1606 | ||
| AF282251.1 | - | 7 | 1297 | |||
| AK081011.1 | - | 6 | 1050 | |||
| AK158641.1 | - | 6 | 977 | |||
| AK158892.1 | - | 6 | 1587 | |||
| AK163648.1 | - | 6 | 1078 | |||
| BC057129.1 | - | 6 | 1594 | 6811387 | ||
| BC058963.1 | - | 6 | 1594 | 6811387 | ||

Caln1
Genomic Map